missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TACSTD2/TROP2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | TACSTD2/TROP2 |
|---|---|
| Concentration | 750 μg/mL |
| Applications | Western Blot, Immunohistochemistry, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17689616
|
Proteintech
27360-1-AP-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17679616
|
Proteintech
27360-1-AP-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TACSTD2/TROP2 Polyclonal antibody apecifically detetects TACSTD2/TROP2 antigen in Human Samples. It is validated for Western Blot, Immunohistochemistry, ELISA.Specifications
| TACSTD2/TROP2 | |
| Western Blot, Immunohistochemistry, ELISA | |
| Unconjugated | |
| Rabbit | |
| PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
| 4070 | |
| TACSTD2/TROP2 fusion protein Ag25769 | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 750 μg/mL | |
| Polyclonal | |
| Liquid | |
| Human | |
| BC009409 | |
| TACSTD2 | |
| IgG | |
| Antigen affinity purification |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title