missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TACR2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25270-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene belongs to a family of genes that function as receptors for tachykinins. Receptor affinities are specified by variations in the 5-end of the sequence. The receptors belonging to this family are characterized by interactions with G proteins and 7 hydrophobic transmembrane regions. This gene encodes the receptor for the tachykinin neuropeptide substance K, also referred to as neurokinin A.Specifications
TACR2 | |
Polyclonal | |
Unconjugated | |
TACR2 | |
Neurokinin A receptor, NK 2 receptor, NK 2R, NK2R, NKNAR, SKR, Substance K receptor, TAC2R, tachykinin receptor 2, TACR2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
6865 | |
-20°C | |
Liquid |
Flow Cytometry, Western Blot | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P21452 | |
TACR2 | |
Peptide | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |