missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TACR2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | TACR2 |
---|---|
Concentration | 0.35 mg/mL |
Applications | Flow Cytometry, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16803855
|
Proteintech
25270-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 17-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16893845
|
Proteintech
25270-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 17-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene belongs to a family of genes that function as receptors for tachykinins. Receptor affinities are specified by variations in the 5-end of the sequence. The receptors belonging to this family are characterized by interactions with G proteins and 7 hydrophobic transmembrane regions. This gene encodes the receptor for the tachykinin neuropeptide substance K, also referred to as neurokinin A.Specifications
TACR2 | |
Flow Cytometry, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P21452 | |
6865 | |
Peptide | |
Primary | |
-20°C | |
TACR2 |
0.35 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Neurokinin A receptor, NK 2 receptor, NK 2R, NK2R, NKNAR, SKR, Substance K receptor, TAC2R, tachykinin receptor 2, TACR2 | |
TACR2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |