Learn More
Eppendorf™ Swing Bucket Rotor for Benchtop Centrifuge
Product Code. 10580152 Shop All Eppendorf Products
Change view
Click to view available options
For Use With (Equipment):
A-4-38
A-4-44
A-4-62
A-4-62-MTP
A-4-81
A-4-81-MTP
A-8-17
Includes:
4 Hangers for 2 x 50 mL Falcon-G
4 MTP buckets
4 x 100mL rectangular buckets
4 x 250mL rectangular buckets
4 x 500mL rectangular buckets
4 x 85mL buckets
8 x 15mL buckets
Unit Size:
Each
Product Code. For Use With (Equipment) Includes unitSize
10580152 A-4-62 4 x 250mL rectangular buckets Each
10258211 A-4-38 4 x 85mL buckets Each
10054271 A-4-44 4 Hangers for 2 x 50 mL Falcon-G Each
10794563 A-4-81-MTP 4 MTP buckets Each
10120052 A-4-81 4 x 500mL rectangular buckets Each
10282811 A-4-44 4 x 100mL rectangular buckets Each
10084161 A-8-17 8 x 15mL buckets Each
10394792 A-4-62-MTP 4 MTP buckets Each
8 options
Product Code. 10580152
Brand: Eppendorf™ 5810709008
Use these rotors for a wide range of molecular applications. Eppendorf™ Swing Bucket Rotors are designed for use with Eppendorf benchtop centrifuges.
  • Accommodates tubes and plates
  • Aerosol-tight caps for safe centrifugation of hazardous samples optionally available
  • Rotor life time: max. 40,000 cycles
  • Rotor, buckets, caps and adapters are autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Type Swinging Bucket Rotor
For Use With (Equipment) A-4-62
Includes 4 x 250mL rectangular buckets
No. of Places 4