Learn More
Memmert™ Support Basket with Perforated Mounting Shelf
Product Code. p-7220966 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
WTB15
WTB24
WTB35
WTB50
Unit Size:
Each
Product Code. 17120315
Brand: Memmert™ WTB15B48814
Only null left
Add to basket
Add to basket
Support basket with perforated mounting shelf for conical flasks clips
  • Use of test tube racks not possible
  • Has grid perforations to take clips for conical flasks
Specifications
For Use With (Equipment) WTB15
Type Support Basket With Perforated Mounting Shelf (Stainless Steel)
Material Stainless Steel