Learn More
Cytiva™ Superose™ 6 Increase Columns
Product Code. 15383224 Shop All Cytiva Products
Change view
Click to view available options
Diameter (Metric) Inner:
3.2 mm
5 mm
10 mm
Height (Metric):
150 mm
300 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner Height (Metric) Unit Size
15383224 5 mm 150 mm Each
15377653 10 mm 300 mm Each
15303234 3.2 mm 300 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15383224 Supplier Cytiva™ Supplier No. 29091597
  • ONLINE EXCLUSIVE PRICE
    30055.32 NOK
    28608.00 NOK / Pack of 1
    Save 1447.32 NOK
    4% Off
 Restricted Item
In Stock
High-resolution size-exclusion chromatography for studying large proteins and complexes. Cytiva Lifescience™ Superose™ 6 Increase Columns delivers high quality in size exclusion chromatography (SEC).
Superose™ 6 Increase pre-packed columns deliver the quality and high resolution in size exclusion chromatography (SEC) — even for large proteins and aggregates.
Increased performance
  • Higher resolution than traditional Superose 6 columns
  • Wide fractionation range (Mr from 5,000 to 5,000,000)
  • Withstands high-pH cleaning
TRUSTED_SUSTAINABILITY
Specifications
Product Type Superose 6 Increase 5/150 GL
Diameter (Metric) Inner 5 mm
Height (Metric) 150 mm
Flow Rate 0.3mL/min.
Material Borosilicate glass
Ligand Type Cross-linked agarose
Max. Pressure 435 psi
pH Range 3 to 12
Particle Size 8.6μm
Support Type Polyethylene (PE) filter
Content And Storage 4° to 30° in 20% ethanol
Column Type Gel Filtration Column
Show More Show Less
compliance-icons
Product Identifier
  • Superose 6 Increase 5/150 GL
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contain: ethanol