Learn More
Cytiva Superdex 75 Increase Prepacked Column, 3.2/300
Product Code. 15599244 Shop All Cytiva Products
Change view
Click to view available options
Volume (Metric) Bed:
∼2.4 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Bed Unit Size
15599244 ∼2.4 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15599244 Supplier Cytiva Supplier No. 29148723
  • ONLINE EXCLUSIVE PRICE
    32058.60 NOK
    30515.00 NOK / Pack of 1
    Save 1543.60 NOK
    4% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Use this column for rapid purity check and screening of proteins with molecular weights between Mr 3,000 and 70,000, such as recombinant tagged proteins. Cytiva Superdex 75 Increase Prepacked Columns are ideal when time is limited and low buffer and sample consumption is important.
  • Up to 50% higher resolution compared with predecessor Superdex 75, for improved purity and analysis results
  • Runtime reduced down to one third compared with predecessor Superdex 75, with maintained resolution for results faster
  • Versatile use in both preparative and analytical applications, especially with recombinant tagged proteins
  • Tolerance to repeated harsh cleaning procedures at high pH ensures long column lifetime and minimizes carryover
  • Higher lot-to-lot consistency compared with predecessor Superdex 75
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) High resolution size exclusion chromatography (SEC) in micro scale (4 to 50μL sample volume)
Volume (Metric) Bed ∼2.4 mL
Content And Storage 4°C to 30°C