Learn More
Cytiva Superdex 75 Increase Prepacked Column, 10/300 GL
Product Code. 15579244 Shop All Cytiva Products
Change view
Click to view available options
Volume (Metric) Bed:
∼24 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Bed Unit Size
15579244 ∼24 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15579244 Supplier Cytiva Supplier No. 29148721
  • ONLINE EXCLUSIVE PRICE
    29815.00 NOK
    28608.00 NOK / Pack of 1
    Save 1207.00 NOK
    4% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Use these columns in preparative purification as well as for characterization and analysis of proteins with molecular weights between Mr 3,000 and 70,000, such as recombinant tagged proteins. Cytiva HisTrap™ excel Prepacked Columns feature tolerance to repeated harsh cleaning procedures at high pH ensures long column lifetime and minimizes carryover.
  • Up to 50% higher resolution compared with predecessor Superdex 75, for improved purity and analysis results
  • Runtime reduced down to one third compared with predecessor Superdex 75, with maintained resolution for results faster
  • Versatile use in both preparative and analytical applications, especially with recombinant tagged proteins
  • Tolerance to repeated harsh cleaning procedures at high pH ensures long column lifetime and minimizes carryover
  • Higher lot-to-lot consistency compared with predecessor Superdex 75
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) High resolution size exclusion chromatography (SEC) in small scale (25 to 500μL sample volume)
Volume (Metric) Bed ∼24 mL
Content And Storage 4°C to 30°C
compliance-icons
Product Identifier
  • Superdex 75 Increase, 10/300 GL
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol