Learn More
Thermo Scientific™ Sulfo-SBED Biotin Label Transfer Reagent, No-Weigh™ Format
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ Pierce Sulfo-SBED Biotin Label Transfer Reagent is a multifunctional reagent for labeling a purified protein and then covalently transferring the attached biotin tag onto specific interactors of that protein.
Highlights:
- Sulfo-NHS ester reacts with primary amines at pH 7-9
- Aryl azide conjugates randomly when activated with UV light
- Cleavable disulfide bond allows biotin label transfer
- Discover protein interactions without radiolabeling
Includes:
No-Weigh Sulfo-SBED Label Transfer Reagent, 8 × 1mg; BupH Phosphate Buffered Saline, 1 Pk. (makes 500mL); Label Transfer Buffer (20X), 200mL (makes 4L); Streptavidin-HRP Conjugate, 0.1mg; No-Weigh Dithiothreitol (DTT), 8 × 7.7mg; Slide-A-Lyzer MINI Dialysis Units plus Float, 10K MWCO, 10-100μL, 10 units/Pk.
Specifications
Specifications
| Description | Pierce Sulfo-SBED Biotin Label Transfer Reagent, No-Weigh Format |
| Formulation | Sulfo-N-hydroxysuccinimidyl-2-(6-[biotinamido]-2-(p-azido benzamido)-hexanoamido) ethyl-1,3'-dithioproprionate; powder solid |
| Quantity | 8 x 1 mg |
| Sufficient For | 18mL of typical 1X (0.5mM) reaction solution (total), or 8 separate uses |
| Molecular Weight (g/mol) | 879.98 |
| Solubility | Water |
| Chemical Reactivity | Amine-to-Nonselective |