Learn More
Memmert™ Subframe For Stacking Vacuum Oven And Pump Module, Total Height 1650 mm, For Vo101
Product Code. 16384517 Shop All Memmert Products
Change view
Click to view available options
Depth (Metric):
533 mm
Unit Size:
Each
Product Code. Depth (Metric) unitSize
16384517 533 mm Each
1 options
Product Code. 16384517
Brand: Memmert™ VO101E02037
Only null left
Specifications
For Use With (Equipment) Model 500
Type Tubular
Material Black Enamel Coated Stainless Steel
Width (Metric) 689 mm
Depth (Metric) 533 mm