Learn More
Memmert™ Subframe on Castors for UN55plus
Product Code. 17540316 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UN55plus Oven
Unit Size:
Each
Product Code. 17540316
Brand: Memmert™ UN55plusB29748
Only null left
Add to basket
Add to basket
Subframe on wheels for Memmert Ovens.
  • On Castors
  • Height: 660 mm
Specifications
Description On Wheels
For Use With (Equipment) UN55plus Oven
Type Subframe
Height (Metric) 660 mm