Learn More
Invitrogen™ eBioscience™ Streptavidin FITC Conjugate
Product Code. 15506536
Change view
Click to view available options
Quantity:
250 μg
Unit Size:
250µg
Product Code. Quantity unitSize
15506536 250 μg 250µg
1 options
Contains Azide
Product Code. 15506536
Brand: Invitrogen™ 11431787
Contains Azide
Commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry. Streptavidin binds to biotin with high affinity.
TRUSTED_SUSTAINABILITY
Specifications
Concentration 0.5 mg/mL
Conjugate FITC (Fluorescein)
Quantity 250 μg
Form Liquid
Product Line eBioscience
Product Type Streptavidin Conjugate (Fluorescent)
Target Molecule Biotin
Label or Dye FITC (Fluorescein)