Learn More
Invitrogen™ Streptavidin - Cy5
Description
Reactivity: CY5-SA reacts with biotin.
Specifications
Specifications
| Concentration | 1 mg/mL |
| Conjugate | Cy5 |
| Quantity | 1 mL |
| Content And Storage | Store in refrigerator (2–8°C). |
| Shipping Condition | Wet Ice |
| Form | Liquid |
| Product Line | Cy |
| Product Type | Streptavidin Conjugate (fluorescent) |