Learn More
IBA Lifesciences Strep-Tactin™XT Superflow™ Cartridge
Product Code. 15728207 Shop All IBA Lifesciences Products
Change view
Click to view available options
Capacity (Metric):
1 x 1 mL
1 x 5 mL
Unit Size:
Each
Product Code. Capacity (Metric) unitSize
15728207 1 x 5 mL Each
15583077 1 x 1 mL Each
2 options
Product Code. 15728207
Brand: IBA Lifesciences 24022001
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Strep-Tactin™XT Superflow™ cartridge is a HPLC/FPLC cartridge to purify Strep-tag™II or Twin-Strep-tag™ fusion proteins
  • Strep-Tactin™XT Superflow™ cartridges contain a 6% agarose coupled with the streptavidin variant Strep-Tactin™XT and are applicable for purification of Strep-tag™II or Twin-Strep-tag™ fusion proteins with HPLC/FPLC devices like Äkta™ systems (Cytiva).
  • In comparison to Strep-Tactin™ Superflow™, Strep-Tactin™XT Superflow™ has a higher affinity for Strep-tag™ fusion proteins, which leads to an improved immobilization and purification.
  • In addition, Strep-Tactin™XT Superflow™ allows the application of denaturing agents such as urea during the purification procedure.
  • As with any other Strep-Tactin™XT resins, the unspecific protein binding is extremely low, which results in an outstanding purity of the target protein. 
Specifications
Binding Capacity 5.7 mg protein/ mL resin
Capacity (Metric) 1 x 5 mL
Quantity 1 Cartridge
For Use With (Application) Protein Purification
Sufficient For Chromatpgraphy workstations with 10 to 32 connections
For Research Use Only.