Learn More
IBA GmbH™ Strep-Tactin™XT Starter Kit
Shop All IBA Lifesciences Products
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Description
- The Strep-Tactin™XT Starter Kit contains all essential reagents required for purification and detection of Strep-tagII™ and Twin-Strep-tag™ proteins.
- The kit contains all reagents sufficient for 8 applications
- 1x 1 mL Strep-Tactin™XT Superflow™ gravity flow column
- Strep-Tactin™/ Strep-Tactin™XT Wash Buffer (1x Buffer W)
- Strep-Tactin™XT Elution Buffer with Biotin for displacing the Twin-Strep-tag™ protein from the column (1x Buffer BXT)
- Strep-Tactin™ horse radish peroxidase (HRP) conjugate for Western blot detection
Specifications
Specifications
Kit Contents | Gravity flow column, wash buffer, elution buffer |
Quantity | 1 Kit |
Product Content Correction
Product Title
IBA GmbH™ Strep-Tactin™XT Starter Kit >