Learn More
IBA Lifesciences Strep-Tactin™ Superflow™ High Capacity Column
Shop All IBA Lifesciences Products
Click to view available options
Quantity:
1 mL
5 mL
5 x 0.2 mL
Unit Size:
1mL
Pack of 5
Description
- Strep-Tactin™ Superflow™ high capacity columns contain a 6% agarose coupled with the streptavidin variant Strep-Tactin™ and are applicable for purification of Strep-tag™II or Twin-Strep-tag™ fusion proteins via gravity flow.
- In comparison to Strep-Tactin™ Superflow™, Strep-Tactin™ Superflow™ high capacity has a higher density of immobilized Strep-Tactin™, which leads to a 3- to 5-fold higher yield of purified protein.
- To reduce the waiting period during sample and buffer application, the use of the application aid WET FRED is recommended.
Specifications
Specifications
| Binding Capacity | 150 to 500 nmoL |
| Capacity (Metric) | 1 mL |
| Support Type | Superflow 6 |
| Quantity | 5 x 0.2 mL |
| For Use With (Application) | One-Step Purification of Recombinant Strep-Tag Proteins |
| Type | Gravity Flow Column |
For Research Use Only.