Learn More
IBA Lifesciences Strep-Tactin™ Sepharose™ Resin
Product Code. 12723074 Shop All IBA Lifesciences Products
Change view
Click to view available options
Quantity:
4 mL
20 mL
50 mL
1000 mL
Unit Size:
1 L
20mL
Each
Product Code. Quantity unitSize
12723074 4 mL Each
12846436 20 mL 20mL
10049889 50 mL Each
12876436 1000 mL 1 L
4 options
Product Code. 12723074
Brand: IBA Lifesciences 21201002
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Strep-Tactin™ Sepharose™ resin is a 50% suspension to purify Strep-tag™II or Twin-Strep-tag™ fusion proteins
  • Strep-Tactin™ Sepharose™ is a 4% agarose coupled with the streptavidin variant Strep-Tactin™ and applicable for purification of Strep-tag™II or Twin-Strep-tag™ fusion proteins.
  • Due to the low concentrated agarose, it is particularly suitable for large proteins, especially if the affinity to Strep-Tactin™XT 4Flow™ is too high.
  • The wide-meshed polymer allows for the simple attachment of larger proteins to the stationary phase.
  • The suspension can be used for packing of gravity flow columns since the resin is optimized for column affinity chromatography, rather than for batch purification.
  • To pack a column with 1 ml resin bed volume, 2 ml 50% suspension is necessary.
Please note that Strep-Tactin™ Sepharose™ is not compatible with HPLC/FPLC due to the low pressure stability.
Specifications
Product Type Strep-Tactin Sepharose Suspension
Quantity 4 mL
Content And Storage 4°C
For Use With (Application) Used Preferentially for Gravity Flow Purification of Recombinant Strep-Tag Proteins
Matrix Strep-Tactin Sepharose
For Research Use Only.