Learn More
IBA Lifesciences Strep-Tactin™ protein as lyophilized powder
Shop All IBA Lifesciences Products
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Description
Strep-Tactin™ is a streptavidin variant, which can be utilized for coating of different surfaces. It is optimized for the binding of Strep-tag™II and Twin-Strep-tag™ fusion proteins but still maintains the ability to capture biotinylated targets. Therefore, it can be used for assay development and protein interaction studies, including experiments such as proximity-dependent biotin identification (BioID).
Specifications
Specifications
| Binding Capacity | >50 nmol/mg |
| Form | Lyophilized |
| Quantity | 5 mg |