Learn More
IBA Lifesciences Strep-Tactin™ protein as lyophilized powder
GREENER_CHOICE
Product Code. 17079133 Shop All IBA Lifesciences Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17079133 1 mg 1mg
17171921 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17079133 Supplier IBA Lifesciences Supplier No. 21204001
Strep-Tactin™ protein as lyophilized powder can be used for reversible immobilization of biotinylated analytes or Strep-tag™II and Twin-Strep-tag™ fusion proteins
Strep-Tactin™ is a streptavidin variant, which can be utilized for coating of different surfaces. It is optimized for the binding of Strep-tag™II and Twin-Strep-tag™ fusion proteins but still maintains the ability to capture biotinylated targets. Therefore, it can be used for assay development and protein interaction studies, including experiments such as proximity-dependent biotin identification (BioID).
Specifications
Binding Capacity >50 nmol/mg
Form Liquid
Quantity 1 mg