missing translation for 'onlineSavingsMsg'
Learn More

STK3 Mouse anti-Human, Mouse, Rat, Clone: 1C11A4, Proteintech

Mouse Monoclonal Antibody

1710.33 NOK - 4836.10 NOK

Specifications

Antigen STK3
Clone 1C11A4
Concentration 1 mg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16808705
View Documents
Proteintech
66637-1-IG-20UL
20 μL
1710.33 NOK
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16898695
View Documents
Proteintech
66637-1-IG-150UL
150 μL
4836.10 NOK
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Required for DNA recombination, repair and replication. The activity of RP-A is mediated by single-stranded DNA binding and protein interactions. Required for the efficient recruitment of the DNA double-strand break repair factor RAD51 to chromatin in response to DNA damage.
Specifications

Specifications

STK3
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
KRS1, MST 2, MST2, STE20 like kinase MST2, STK3
Stk3
IgG2a
Protein A
Antibody
1C11A4
Western Blot
Unconjugated
Mouse
Mouse, Rat, Human
O54748, Q13188, Q9JI10
56274, 65189, 6788
STK3 Fusion Protein Ag2741
Primary
-20°C
STK3
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.