Learn More
STK3 Mouse anti-Human, Mouse, Rat, Clone: 1C11A4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66637-1-IG-20UL
This item is not returnable.
View return policy
Description
Required for DNA recombination, repair and replication. The activity of RP-A is mediated by single-stranded DNA binding and protein interactions. Required for the efficient recruitment of the DNA double-strand break repair factor RAD51 to chromatin in response to DNA damage.Specifications
| STK3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O54748, Q13188, Q9JI10 | |
| Stk3 | |
| STK3 Fusion Protein Ag2741 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| 1C11A4 | |
| Unconjugated | |
| STK3 | |
| KRS1, MST 2, MST2, STE20 like kinase MST2, STK3 | |
| Mouse | |
| Protein A | |
| RUO | |
| 56274, 65189, 6788 | |
| -20°C | |
| Liquid |