Learn More
Fisherbrand™ Sterile Graduated Transfer Pipets
Product Code. 13489108 Shop All Fisherbrand Products
Change view
Click to view available options
Volume (Metric):
1 mL
3 mL
Length (Metric):
150 mm
155 mm
Volume (Metric) Dispense:
33 μL
40 μL
Packaging Type:
25 inner packs of 20
50 inner packs of 10
Individually Wrapped
Unit Size:
Pack of 500
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Length (Metric) Volume (Metric) Dispense Packaging Type unitSize
13489108 1 mL 150 mm 33 μL Individually Wrapped Pack of 500
13419118 1 mL 150 mm 33 μL 50 inner packs of 10 Pack of 500
13469108 3 mL 155 mm 40 μL Individually Wrapped Pack of 500
13409118 3 mL 155 mm 40 μL 25 inner packs of 20 Pack of 500
13479108 3 mL 155 mm 40 μL 50 inner packs of 10 Pack of 500
13429118 1 mL 150 mm 33 μL 25 inner packs of 20 Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 13489108
Spotlight Offers
Use promo code 28036
View all promotions
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
Made up of low density polyethylene.
  • Transparent in color
  • Contains graduations
  • Sterile
Specifications
Material LDPE
Drops/mL 30
Graduated Yes
Graduations 1 mL
Quantity 500/Pk.
Volume (Metric) Dispense 33 μL
Packaging Type Individually Wrapped
Volume (Metric) 1 mL
Length (Metric) 150 mm
Type Transfer Pipet