Learn More
Fisherbrand™ Sterile Graduated Transfer Pipets
Product Code. 13469108 Shop All Fisherbrand Products
Change view
Click to view available options
Volume (Metric):
1 mL
3 mL
Length (Metric):
150 mm
155 mm
Volume (Metric) Dispense:
33 μL
40 μL
Packaging Type:
25 inner packs of 20
50 inner packs of 10
Individually Wrapped
Unit Size:
Pack of 500
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Length (Metric) Volume (Metric) Dispense Packaging Type unitSize
13469108 3 mL 155 mm 40 μL Individually Wrapped Pack of 500
13489108 1 mL 150 mm 33 μL Individually Wrapped Pack of 500
13419118 1 mL 150 mm 33 μL 50 inner packs of 10 Pack of 500
13409118 3 mL 155 mm 40 μL 25 inner packs of 20 Pack of 500
13479108 3 mL 155 mm 40 μL 50 inner packs of 10 Pack of 500
13429118 1 mL 150 mm 33 μL 25 inner packs of 20 Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 13469108
Add to basket
Edge
Aggiungi al carrello
Made up of low density polyethylene.
  • Transparent in color
  • Contains graduations
  • Sterile
Specifica
Material LDPE
Drops/mL 25
Graduated Yes
Graduations 0.5 mL
Quantity 500/Pk.
Volume (Metric) Dispense 40 μL
Packaging Type Individually Wrapped
Volume (Metric) 3 mL
Length (Metric) 155 mm
Type Transfer Pipet
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.