Learn More
Fisherbrand™ Sterile Disposable Standard Serological Pipets
Product Code. 11899660 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Volume (Metric):
1 mL
2 mL
5 mL
10 mL
25 mL
50 mL
Graduations:
0.01 mL
0.1 mL
0.10 mL
0.2 mL
0.5 mL
Tip Style:
Standard
Tolerance:
±0.02 mL
±0.04 mL
±0.1 mL
±0.2 mL
±0.5 mL
±1 mL
Unit Size:
Each
Pack of 1000
Pack of 200
Pack of 500
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Graduations Tip Style Tolerance Unit Size
11899660 5 mL 0.10 mL Standard ±0.1 mL Pack of 500
11879660 1 mL 0.01 mL Standard ±0.02 mL Pack of 1000
11889660 2 mL 0.01 mL Standard ±0.04 mL Pack of 500
11809670 10 mL 0.1 mL Standard ±0.2 mL Pack of 500
11819670 25 mL 0.2 mL Standard ±0.5 mL Pack of 200
10421645 50 mL 0.5 mL Standard ±1 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11899660
  • ONLINE EXCLUSIVE PRICE
    1290.00 NOK
    902.68 NOK / Pack of 500
    Save 387.32 NOK
    30% Off
Estimated Shipment: 01-12-2026
Add to basket
Add to basket
Polystyrene pipet with negative graduations on all sizes
Fisherbrand™ Sterile Disposable Standard Serological Pipets feature a standard tip. Polystyrene construction, with negative graduations on all sizes. Certified ASTM E934, ASTM E1380 compliant.
  • Ascending and descending graduations on all but the 1 and 2mL sizes
  • Sterile and nonwettable pipets
  • Offer a tolerance of ±2% at full volume and have plug-in mouth end
  • Without magnifier stripe
  • Recyclable outer packaging
  • Bulk packed
  • Each pipet lot is tested for sterility, accuracy and endotoxin level
Certifications
Certified nonpyrogenic with an endotoxin level less than 0.5EU/mL following USP XXII Bacterial Endotoxins Test (85); DNase/RNase Free; BSE/TSE Free
Compliance
Designed from ASTM™ E 934; conform to ASTM E 1380 for color coding. Calibrated To Deliver/Blow Out
TRUSTED_SUSTAINABILITY
Specifications
Calibration To Deliver/Blow Out
Certifications/Compliance ASTM E934, ASTM E1380; DNase/RNase Free; BSE/TSE Free
Material Polystyrene
Sterility Sterile
Graduated Yes
Graduations 0.10 mL
Negative Graduations 2 mL
Quantity 50/Pk., 500/Cs.
Tip Style Standard
Tolerance ±0.1 mL
Packaging Type Bulk
Volume (English) 0.16 oz.
Color Blue
Volume (Metric) 5 mL
Type Serological Pipet