Learn More
R&L Enterprises™ Steel Wire Gauze
Product Code. 12958950 Shop All Fisherbrand Products
Change view
Click to view available options
Dimensions (L x W):
125 x 125 mm
150 x 150 mm
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
12958950 125 x 125 mm Pack of 10
12968950 150 x 150 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12958950
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
Provides excellent support to beakers and other glassware during heating on ring stands and tripods. R&L Enterprises™ Steel Wire Gauze is a flat square manufactured from steel mesh.
  • Non-fraying edges
  • Durable and long-lasting
  • Excellent heat distribution
Specifications
Ceramic Center Ceramic Center
Dimensions (L x W) 125 x 125 mm
Edges Non-Fraying
Quantity 10 Pack
Shape Square
Length (Metric) 125 mm
Material Steel
Type Wire Gauze
Width (Metric) 125 mm