Learn More
Fisherbrand™ Stainless Steel Straight Tongs
Product Code. 11593492 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Length (Metric):
150 mm
200 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
11593492 150 mm Each
11503502 200 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11593492
  • ONLINE EXCLUSIVE PRICE
    68.70 NOK
    61.21 NOK / Pack of 1
    Save 7.49 NOK
    10% Off
Estimated Shipment: 08-01-2027
Add to basket
Add to basket
Get maximum results using laboratory tongs in high temperature chemical reactions. Made of durable stainless steel, these Clifton™ Stainless Steel Straight Tongs ensure precise control of grasping, lifting, turning, and transferring heat-resistant and other materials in a laboratory environment.
  • Laboratory straight tongs
  • Stainless steel 304 spec 18/8 non-magnetic, bright finish
  • Designed for corrosion resistance and durability ideal in laboratory routines
  • Laboratory metalware consumables
  • Finest in corrosion-resistant 304 stainless steel
  • Provides corrosion resistance that prevents product contamination
Specifications
Product Type Crucible Tongs
Quantity 1 Ea.
Length (Metric) 150 mm
Material Steel