Learn More
STAT3 / Signal Transducer and Activator of Transcription 3 Mouse, Clone: PCRP-STAT3-2F12, enQuireBio™
Mouse Monoclonal Antibody
Brand:  EnquireBio 6774-MSM1-P1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15950928
  • 5275.00 NOK / 100µg
Estimated Shipment: 03-10-2024
Specifications
STAT3 / Signal Transducer and Activator of Transcription 3
Cytoplasmic and Nuclear
PCRP-STAT3-2F12
Unconjugated
Mouse
88kDa
Protein A/G purified from Bioreactor concentrate.
RUO
6774
IgG2a
ELISA
Monoclonal
200μg/mL
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide.
Recombinant full-length human STAT3 protein
A431 and Raji cells. Kidney, Brain or Heart
100 μg
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.