missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAT3 / Signal Transducer and Activator of Transcription 3 Mouse, Clone: PCRP-STAT3-2F12, enQuireBio™
Mouse Monoclonal Antibody
2058.63 NOK - 5275.00 NOK
Specifications
Antigen | STAT3 / Signal Transducer and Activator of Transcription 3 |
---|---|
Clone | PCRP-STAT3-2F12 |
Concentration | 200μg/mL |
Applications | ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15950928
|
EnquireBio
6774-MSM1-P1 |
100 μg |
5275.00 NOK
100µg |
Estimated Shipment: 08-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15940928
|
EnquireBio
6774-MSM1-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
STAT3 / Signal Transducer and Activator of Transcription 3 | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant full-length human STAT3 protein | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
88kDa |
PCRP-STAT3-2F12 | |
ELISA | |
Unconjugated | |
RUO | |
6774 | |
IgG2a | |
Protein A/G purified from Bioreactor concentrate. | |
Cytoplasmic and Nuclear | |
A431 and Raji cells. Kidney, Brain or Heart |