missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAT3 / Signal Transducer and Activator of Transcription 3 Mouse, Clone: PCRP-STAT3-2F12, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 6774-MSM1-P0
Additional Details : Weight : 0.01000kg
Specifications
STAT3 / Signal Transducer and Activator of Transcription 3 | |
Cytoplasmic and Nuclear | |
PCRP-STAT3-2F12 | |
Unconjugated | |
Mouse | |
88kDa | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
6774 | |
IgG2a |
ELISA | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant full-length human STAT3 protein | |
A431 and Raji cells. Kidney, Brain or Heart | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |