Learn More
Eppendorf Starter Set 100 μL 384-format: Eppendorf Xplorer™ Plus Move It™
Shop All Eppendorf Products
Click to view available options
Number of Channels:
8
12
Unit Size:
1 set
Description
- 4-, 6-, 8- and 12-channel air-cushion pipettes with variable volume and adjustable tip spacing option
- Available as mechanical (Research plus) and electronic (Xplorer plus) adjustable tip spacing multi-channel pipette
- For plates (384-, 96-, 48-, 24- and 12-well), tubes, agarose gels and more (depending on pipette variant)
- Tip spacing freely selectable from 4.5 to 33 mm, depending on pipette variant
- Format limiter for quick switches backwards and forwards between source and destination formats
- Manual adjustment knob for smooth vibration-free format switches and liquid transfers without dripping
- Tubeless system for enhanced reliability and safety
- Autoclavable for user and sample safety (Research plus fully, and Xplorer plus lower part only)
- 360° rotatable pipette head for comfortable readability of display and fast identification of parameters
- Optimally balanced pipette - reduces hand fatigue for stress-free operation
Specifications
Specifications
| Certifications/Compliance | Lot-Specific Certificate |
| Compatible Tips | epT.I.P.S.™, epT.I.P.S.™ 384 |
| Autoclavable | Autoclavable |
| Color | Light Yellow |
| Disposable | Single-use |
| Number of Channels | 8 |
| Sterility | Sterile |
| Type | Manual Pipettes |
| Volume (Metric) | 5 to 100 μL |
| For Use With (Equipment) | Air-cushion System |
| Show More |