missing translation for 'onlineSavingsMsg'
Learn More

STARS/ABRA Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 22673-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16892035

  • 1611.84 NOK / 20µL
Estimated Shipment: 22-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Acts as an activator of serum response factor (SRF)-dependent transcription possibly by inducing nuclear translocation of MKL1 or MKL2 and through a mechanism requiring Rho-actin signaling.
Specifications

Specifications

STARS/ABRA
Polyclonal
Unconjugated
ABRA
ABRA, STARS
Rabbit
Antigen Affinity Chromatography
RUO
137735, 223513
-20°C
Liquid
Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot
0.26 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8BUZ1, Q8N0Z2
ABRA
STARS/ABRA Fusion Protein Ag18456
20 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers