missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STARS/ABRA Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | STARS/ABRA |
---|---|
Concentration | 0.26 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16892035
|
Proteintech
22673-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16882035
|
Proteintech
22673-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Acts as an activator of serum response factor (SRF)-dependent transcription possibly by inducing nuclear translocation of MKL1 or MKL2 and through a mechanism requiring Rho-actin signaling.Specifications
STARS/ABRA | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q8BUZ1, Q8N0Z2 | |
137735, 223513 | |
STARS/ABRA Fusion Protein Ag18456 | |
Primary | |
-20°C | |
ABRA |
0.26 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ABRA, STARS | |
ABRA | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |