Learn More
Ohaus™ Stainless Steel Weight, OIML E2, OH
Single body design weight adjusted to meet or exceed 0IML E2 tolerances
Brand: Ohaus™ 11117005
Additional Details : Weight : 0.20000kg
Description
- Stainless steel construction
- Include a weight case
- Single body design without adjusting cavity
Specifications
Weight, 200g, OIML E2, OH | |
Stainless Steel | |
Meets or exceeds OIML E2 |
OIML E2 Weights | |
200 g |