Learn More
Ohaus™ Stainless Steel Weight, OIML E2, OH
Single body design weight adjusted to meet or exceed 0IML E2 tolerances
1695.00 NOK - 2120.00 NOK
Specifications
Classification | OIML E2 Weights |
---|---|
Material | Stainless Steel |
Tolerance | Meets or exceeds OIML E2 |
Product Code | Brand | Weight (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Weight (Metric) | Price | Quantity & Availability | |||||
15653384
|
Ohaus™
11117003 |
50 g |
1695.00 NOK
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
15663384
|
Ohaus™
11117004 |
100 g |
1970.00 NOK
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
15732854
|
Ohaus™
11117005 |
200 g |
2120.00 NOK
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
Description
- Stainless steel construction
- Include a weight case
- Single body design without adjusting cavity
Specifications
OIML E2 Weights | |
Stainless Steel | |
Meets or exceeds OIML E2 |