Learn More
Fisherbrand™ Stainless Steel Tripods
Shop All Fisherbrand Products
Click to view available options
Height (Metric):
150 mm
210 mm
260 mm
Width (Metric):
120 mm
150 mm
Unit Size:
Each
Specifications
Specifications
| Material | Stainless Steel |
| Height (Metric) | 260 mm |
| Width (Metric) | 150 mm |
| Includes | Plastic Feet |
| Product Type | Tripod |
| Finish | Bright |
| Quantity | 1 Ea. |