Learn More
- Home
- Products
- Filters and Filtration
- Mechanical Sieves
- RETSCH Stainless-Steel Test Sieves, 400 mm Dia., with Stainless-Steel Wire
RETSCH Stainless-Steel Test Sieves, 400 mm Dia., with Stainless-Steel Wire
Product Code. 10799122 Shop All Retsch Products
Change view
Click to view available options
Pore Size:
1 mm
1.12 mm
1.18 mm
1.25 mm
1.4 mm
1.6 mm
1.7 mm
1.8 mm
2 mm
2.24 mm
2.36 mm
2.5 mm
2.8 mm
3.15 mm
3.35 mm
3.55 mm
4 mm
4.5 mm
4.75 mm
5 mm
5.6 mm
6.3 mm
6.7 mm
7.1 mm
8 mm
9 mm
9.5 mm
10 mm
11.2 mm
12.5 mm
13.2 mm
14 mm
16 mm
18 mm
19 mm
20 mm
22.4 mm
25 mm
26.5 mm
28 mm
31.5 mm
35.5 mm
37.5 mm
40 mm
45 mm
50 mm
53 mm
56 mm
63 mm
Unit Size:
Each
| Product Code. | Pore Size | unitSize |
|---|---|---|
| 10799122 | 2.5 mm | Each |
| 10095031 | 1.12 mm | Each |
| 10136121 | 1.18 mm | Each |
| 10406621 | 1.25 mm | Each |
| 10680082 | 1.4 mm | Each |
| 10106221 | 1.6 mm | Each |
| 10630262 | 1.8 mm | Each |
| 10544302 | 10 mm | Each |
| 10416621 | 1.7 mm | Each |
| 10239071 | 11.2 mm | Each |
| 10417201 | 12.5 mm | Each |
| 10545262 | 16 mm | Each |
| 10014911 | 13.2 mm | Each |
| 10126461 | 19 mm | Each |
| 10608761 | 1 mm | Each |
| 10024911 | 14 mm | Each |
| 10650452 | 18 mm | Each |
| 10477191 | 2.24 mm | Each |
| 10487191 | 2.36 mm | Each |
| 10044941 | 2.8 mm | Each |
| 10545452 | 20 mm | Each |
| 10380382 | 25 mm | Each |
| 10340432 | 26.5 mm | Each |
| 10044981 | 22.4 mm | Each |
| 10146651 | 28 mm | Each |
| 10584112 | 2 mm | Each |
| 10186501 | 3.15 mm | Each |
| 10630092 | 3.35 mm | Each |
| 10478151 | 3.55 mm | Each |
| 10005021 | 31.5 mm | Each |
| 10498921 | 35.5 mm | Each |
| 10288591 | 37.5 mm | Each |
| 10278731 | 4.5 mm | Each |
| 10760663 | 45 mm | Each |
| 10004981 | 4.75 mm | Each |
| 10543342 | 4 mm | Each |
| 10458921 | 5.6 mm | Each |
| 10197021 | 50 mm | Each |
| 10155981 | 40 mm | Each |
| 10610292 | 53 mm | Each |
| 10770663 | 56 mm | Each |
| 10116031 | 6.3 mm | Each |
| 10014981 | 5 mm | Each |
| 10720473 | 6.7 mm | Each |
| 10176071 | 7.1 mm | Each |
| 10045051 | 8 mm | Each |
| 10167121 | 63 mm | Each |
| 10436621 | 9.5 mm | Each |
| 10330912 | 9 mm | Each |
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Specifications
Specifications
| Quantity | 1 Each |
| Pore Size | 2.5 mm |
| Type | Test Sieve |
| Certifications/Compliance | ISO 3310/1 |
| For Use With (Equipment) | Vibratory Shaker AS 450 |
| Includes | Certificate indicating Compliance |
| Height (Metric) | 65mm |
| For Use With (Application) | For separation, fractioning, particle size determination |
| Diameter (Metric) Outer | 400 mm |
| Frame Material | Stainless Steel |
| Mesh Material | Stainless Steel |
| Show More Show Less |