Learn More
RETSCH Stainless-Steel Test Sieves, 400 mm Dia., with Stainless-Steel Wire
Product Code. 10146651 Shop All Retsch Products
Change view
Click to view available options
Pore Size:
1 mm
1.12 mm
1.18 mm
1.25 mm
1.4 mm
1.6 mm
1.7 mm
1.8 mm
2 mm
2.24 mm
2.36 mm
2.5 mm
2.8 mm
3.15 mm
3.35 mm
3.55 mm
4 mm
4.5 mm
4.75 mm
5 mm
5.6 mm
6.3 mm
6.7 mm
7.1 mm
8 mm
9 mm
9.5 mm
10 mm
11.2 mm
12.5 mm
13.2 mm
14 mm
16 mm
18 mm
19 mm
20 mm
22.4 mm
25 mm
26.5 mm
28 mm
31.5 mm
35.5 mm
37.5 mm
40 mm
45 mm
50 mm
53 mm
56 mm
63 mm
Unit Size:
Each
Product Code. Pore Size unitSize
10146651 28 mm Each
10095031 1.12 mm Each
10136121 1.18 mm Each
10406621 1.25 mm Each
10680082 1.4 mm Each
10106221 1.6 mm Each
10630262 1.8 mm Each
10544302 10 mm Each
10416621 1.7 mm Each
10239071 11.2 mm Each
10417201 12.5 mm Each
10545262 16 mm Each
10014911 13.2 mm Each
10126461 19 mm Each
10608761 1 mm Each
10024911 14 mm Each
10650452 18 mm Each
10477191 2.24 mm Each
10487191 2.36 mm Each
10799122 2.5 mm Each
10044941 2.8 mm Each
10545452 20 mm Each
10380382 25 mm Each
10340432 26.5 mm Each
10044981 22.4 mm Each
10584112 2 mm Each
10186501 3.15 mm Each
10630092 3.35 mm Each
10478151 3.55 mm Each
10005021 31.5 mm Each
10498921 35.5 mm Each
10288591 37.5 mm Each
10278731 4.5 mm Each
10760663 45 mm Each
10004981 4.75 mm Each
10543342 4 mm Each
10458921 5.6 mm Each
10197021 50 mm Each
10155981 40 mm Each
10610292 53 mm Each
10770663 56 mm Each
10116031 6.3 mm Each
10014981 5 mm Each
10720473 6.7 mm Each
10176071 7.1 mm Each
10045051 8 mm Each
10167121 63 mm Each
10436621 9.5 mm Each
10330912 9 mm Each
49 options
Product Code. 10146651
Brand: RETSCH 60.166.028000
Only null left
Specifications
Quantity 1 Each
Pore Size 28 mm
Type Test Sieve
Certifications/Compliance ISO 3310/1
For Use With (Equipment) Vibratory Shaker AS 450
Includes Certificate indicating Compliance
Height (Metric) 65mm
For Use With (Application) For separation, fractioning, particle size determination
Diameter (Metric) Outer 400 mm
Frame Material Stainless Steel
Mesh Material Stainless Steel