Learn More
- Home
- Products
- Filters and Filtration
- Mechanical Sieves
- RETSCH Stainless-Steel Test Sieves, 400 mm Dia., with Perforated Plates with Round Holes
RETSCH Stainless-Steel Test Sieves, 400 mm Dia., with Perforated Plates with Round Holes
Product Code. 10296262 Shop All Retsch Products
Change view
Click to view available options
Pore Size:
1 mm
1.25 mm
2 mm
2.36 mm
2.5 mm
2.8 mm
3.15 mm
3.35 mm
3.55 mm
4 mm
4.5 mm
4.75 mm
5 mm
5.6 mm
6.3 mm
6.7 mm
7.1 mm
8 mm
9 mm
9.5 mm
10 mm
11.2 mm
12.5 mm
13.2 mm
14 mm
16 mm
18 mm
19 mm
20 mm
22.4 mm
25 mm
26.5 mm
28 mm
31.5 mm
35.5 mm
37.5 mm
40 mm
45 mm
50 mm
53 mm
56 mm
63 mm
71 mm
75 mm
80 mm
90 mm
100 mm
106 mm
112 mm
125 mm
Unit Size:
Each
| Product Code. | Pore Size | unitSize |
|---|---|---|
| 10296262 | 2.8 mm | Each |
| 10307902 | 1.25 mm | Each |
| 10368372 | 106 mm | Each |
| 10154732 | 100 mm | Each |
| 10572863 | 10 mm | Each |
| 10798784 | 11.2 mm | Each |
| 10378372 | 112 mm | Each |
| 10675163 | 12.5 mm | Each |
| 10164732 | 125 mm | Each |
| 10124542 | 13.2 mm | Each |
| 10092792 | 16 mm | Each |
| 10685163 | 14 mm | Each |
| 10645743 | 18 mm | Each |
| 10032822 | 19 mm | Each |
| 10462262 | 2.36 mm | Each |
| 10276652 | 1 mm | Each |
| 10338512 | 2.5 mm | Each |
| 10286362 | 20 mm | Each |
| 10104782 | 22.4 mm | Each |
| 10412272 | 25 mm | Each |
| 10412462 | 26.5 mm | Each |
| 10686123 | 28 mm | Each |
| 10183812 | 2 mm | Each |
| 10739774 | 3.15 mm | Each |
| 10236412 | 3.35 mm | Each |
| 10032832 | 3.55 mm | Each |
| 10473032 | 37.5 mm | Each |
| 10626703 | 4.5 mm | Each |
| 10738794 | 31.5 mm | Each |
| 10799944 | 45 mm | Each |
| 10105022 | 40 mm | Each |
| 10789934 | 4.75 mm | Each |
| 10574593 | 35.5 mm | Each |
| 10625933 | 4 mm | Each |
| 10012692 | 5.6 mm | Each |
| 10206802 | 53 mm | Each |
| 10338182 | 56 mm | Each |
| 10002692 | 5 mm | Each |
| 10012732 | 50 mm | Each |
| 10464182 | 6.3 mm | Each |
| 10474182 | 6.7 mm | Each |
| 10012742 | 63 mm | Each |
| 10348132 | 7.1 mm | Each |
| 10236322 | 71 mm | Each |
| 10296362 | 75 mm | Each |
| 10296742 | 8 mm | Each |
| 10328232 | 80 mm | Each |
| 10494182 | 90 mm | Each |
| 10634593 | 9.5 mm | Each |
| 10768624 | 9 mm | Each |
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Specifications
Specifications
| Quantity | 1 Each |
| Pore Size | 2.8 mm |
| Type | Test Sieve |
| Certifications/Compliance | ISO 3310/2 |
| Includes | Certificate indicating Compliance |
| Height (Metric) | 65mm |
| For Use With (Application) | For separation, fractioning, particle size determination |
| Diameter (Metric) Outer | 400 mm |
| Frame Material | Stainless Steel |
| Mesh Material | Stainless Steel |