Learn More
RETSCH Stainless-Steel Test Sieves, 315 mm Dia., in Pore Size Greater than 10 mm
Product Code. 11702986 Shop All Retsch Products
Change view
Click to view available options
Pore Size:
11.2 mm
12.5 mm
13.2 mm
14 mm
19 mm
20 mm
22.4 mm
25 mm
26.5 mm
28 mm
31.5 mm
35.5 mm
37.5 mm
40 mm
50 mm
53 mm
56 mm
63 mm
71 mm
75 mm
90 mm
100 mm
106 mm
112 mm
125 mm
Unit Size:
Each
Product Code. Pore Size unitSize
11702986 11.2 mm Each
11772986 20 mm Each
11752996 40 mm Each
11732986 14 mm Each
11773006 112 mm Each
11763006 106 mm Each
11723006 75 mm Each
11782996 53 mm Each
11743006 90 mm Each
11782986 22.4 mm Each
11742996 37.5 mm Each
11703006 63 mm Each
11772996 50 mm Each
11783006 125 mm Each
11762986 19 mm Each
11712986 12.5 mm Each
11753006 100 mm Each
11712996 28 mm Each
11702996 26.5 mm Each
11713006 71 mm Each
11732996 35.5 mm Each
11722996 31.5 mm Each
11792996 56 mm Each
11722986 13.2 mm Each
11792986 25 mm Each
25 options
Product Code. 11702986
Brand: RETSCH 60.172.011200
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Quantity 1 Each
Pore Size 11.2 mm
Type Test Sieve
Certifications/Compliance ISO 3310 and ASTM E 11, DIN 3310/2
For Use With (Application) For screening aggregates, separation, fractioning, particle size determination
Diameter (Metric) Outer 315 mm
Frame Material Stainless Steel
Mesh Material Stainless Steel