Learn More
RETSCH Stainless-Steel Test Sieves, 200mm Dia., with Inspection Certificate
Brand:  RETSCH 60.231.000200
1724.25 NOK valid until 2024-12-31
Use promo code "23265" to get your promotional price.
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 15182433
  • 1815.00 NOK / Each
Estimated Shipment: 28-06-2024
Specifications
Stainless Steel
Stainless Steel
1 Each
Test Sieve
50mm
Separation, fractioning, particle size determination
200 mm
200μm
ISO 3310/1 and ASTM E 11