Learn More
- Home
- Products
- Filters and Filtration
- Mechanical Sieves
- RETSCH Stainless-Steel Test Sieves, 200 Dia. x 50 mmH, with Perforated Plates with Square Holes
RETSCH Stainless-Steel Test Sieves, 200 Dia. x 50 mmH, with Perforated Plates with Square Holes
Product Code. 10032772 Shop All Retsch Products
Change view
Click to view available options
Pore Size:
4 mm
4.5 mm
4.75 mm
5 mm
5.6 mm
6.3 mm
6.7 mm
7.1 mm
8 mm
9 mm
9.5 mm
10 mm
11.2 mm
12.5 mm
13.2 mm
14 mm
16 mm
18 mm
19 mm
20 mm
22.4 mm
25 mm
26.5 mm
28 mm
31.5 mm
35.5 mm
37.5 mm
40 mm
45 mm
50 mm
53 mm
56 mm
63 mm
71 mm
75 mm
80 mm
90 mm
100 mm
106 mm
112 mm
125 mm
Unit Size:
Each
| Product Code. | Pore Size | unitSize |
|---|---|---|
| 10032772 | 5.6 mm | Each |
| 10347942 | 100 mm | Each |
| 10534593 | 10 mm | Each |
| 10636513 | 106 mm | Each |
| 10296452 | 11.2 mm | Each |
| 10032692 | 112 mm | Each |
| 10194242 | 13.2 mm | Each |
| 10442842 | 12.5 mm | Each |
| 10626893 | 16 mm | Each |
| 10042692 | 125 mm | Each |
| 10483222 | 14 mm | Each |
| 10484182 | 19 mm | Each |
| 10236602 | 18 mm | Each |
| 10256792 | 22.4 mm | Each |
| 10515363 | 20 mm | Each |
| 10525363 | 25 mm | Each |
| 10226222 | 26.5 mm | Each |
| 10103632 | 28 mm | Each |
| 10022762 | 31.5 mm | Each |
| 10002802 | 37.5 mm | Each |
| 10582863 | 35.5 mm | Each |
| 10377942 | 4.5 mm | Each |
| 10226272 | 40 mm | Each |
| 10749754 | 4.75 mm | Each |
| 10002812 | 45 mm | Each |
| 10135072 | 4 mm | Each |
| 10286312 | 50 mm | Each |
| 10769944 | 53 mm | Each |
| 10193812 | 5 mm | Each |
| 10778624 | 56 mm | Each |
| 10472262 | 6.7 mm | Each |
| 10769924 | 6.3 mm | Each |
| 10665163 | 7.1 mm | Each |
| 10593823 | 63 mm | Each |
| 10114782 | 71 mm | Each |
| 10748794 | 75 mm | Each |
| 10206512 | 80 mm | Each |
| 10422462 | 90 mm | Each |
| 10749774 | 9.5 mm | Each |
| 10082802 | 8 mm | Each |
| 10709934 | 9 mm | Each |
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Specifications
Specifications
| Quantity | 1 Each |
| Pore Size | 5.6 mm |
| Type | Test Sieve |
| Certifications/Compliance | ISO 3310/2 |
| Includes | Certificate indicating Compliance |
| Height (Metric) | 50mm |
| For Use With (Application) | For separation, fractioning, particle size determination |
| Diameter (Metric) Outer | 200 mm |
| Frame Material | Stainless Steel |
| Mesh Material | Stainless Steel |