Learn More
- Home
- Products
- Filters and Filtration
- Mechanical Sieves
- RETSCH Stainless-Steel Test Sieves, 200 Dia. x 50 mmH, with Perforated Plates with Round Holes and Pore Size Greater than 10 mm
RETSCH Stainless-Steel Test Sieves, 200 Dia. x 50 mmH, with Perforated Plates with Round Holes and Pore Size Greater than 10 mm
Product Code. 10022702 Shop All Retsch Products
Change view
Click to view available options
Pore Size:
11.2 mm
12.5 mm
13.2 mm
14 mm
16 mm
18 mm
19 mm
20 mm
22.4 mm
25 mm
26.5 mm
28 mm
31.5 mm
35.5 mm
37.5 mm
40 mm
45 mm
50 mm
53 mm
56 mm
63 mm
71 mm
75 mm
80 mm
90 mm
100 mm
106 mm
112 mm
125 mm
Unit Size:
Each
| Product Code. | Pore Size | unitSize |
|---|---|---|
| 10022702 | 31.5 mm | Each |
| 10124782 | 100 mm | Each |
| 10042822 | 106 mm | Each |
| 10789924 | 11.2 mm | Each |
| 10124732 | 112 mm | Each |
| 10594203 | 12.5 mm | Each |
| 10655743 | 125 mm | Each |
| 10092802 | 14 mm | Each |
| 10348512 | 13.2 mm | Each |
| 10719934 | 16 mm | Each |
| 10206462 | 18 mm | Each |
| 10544593 | 19 mm | Each |
| 10635933 | 20 mm | Each |
| 10216462 | 22.4 mm | Each |
| 10483412 | 25 mm | Each |
| 10022692 | 26.5 mm | Each |
| 10493412 | 28 mm | Each |
| 10082722 | 35.5 mm | Each |
| 10266792 | 40 mm | Each |
| 10082732 | 37.5 mm | Each |
| 10092732 | 45 mm | Each |
| 10032762 | 50 mm | Each |
| 10348322 | 56 mm | Each |
| 10592863 | 53 mm | Each |
| 10695163 | 71 mm | Each |
| 10042762 | 63 mm | Each |
| 10444762 | 75 mm | Each |
| 10012812 | 80 mm | Each |
| 10144682 | 90 mm | Each |
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Specifications
Specifications
| Quantity | 1 Each |
| Pore Size | 31.5 mm |
| Type | Test Sieve |
| Certifications/Compliance | ISO 3310/2 |
| Includes | Certificate indicating Compliance |
| Height (Metric) | 50mm |
| For Use With (Application) | For separation, fractioning, particle size determination |
| Diameter (Metric) Outer | 200 mm |
| Frame Material | Stainless Steel |
| Mesh Material | Stainless Steel |