Learn More
RETSCH Stainless-Steel Test Sieves, 200 Dia. x 50 mmH, with Perforated Plates with Round Holes and Pore Size Greater than 10 mm
Product Code. 10032762 Shop All Retsch Products
Change view
Click to view available options
Pore Size:
11.2 mm
12.5 mm
13.2 mm
14 mm
16 mm
18 mm
19 mm
20 mm
22.4 mm
25 mm
26.5 mm
28 mm
31.5 mm
35.5 mm
37.5 mm
40 mm
45 mm
50 mm
53 mm
56 mm
63 mm
71 mm
75 mm
80 mm
90 mm
100 mm
106 mm
112 mm
125 mm
Unit Size:
Each
Product Code. Pore Size unitSize
10032762 50 mm Each
10124782 100 mm Each
10042822 106 mm Each
10789924 11.2 mm Each
10124732 112 mm Each
10594203 12.5 mm Each
10655743 125 mm Each
10092802 14 mm Each
10348512 13.2 mm Each
10719934 16 mm Each
10206462 18 mm Each
10544593 19 mm Each
10635933 20 mm Each
10216462 22.4 mm Each
10483412 25 mm Each
10022692 26.5 mm Each
10022702 31.5 mm Each
10493412 28 mm Each
10082722 35.5 mm Each
10266792 40 mm Each
10082732 37.5 mm Each
10092732 45 mm Each
10348322 56 mm Each
10592863 53 mm Each
10695163 71 mm Each
10042762 63 mm Each
10444762 75 mm Each
10012812 80 mm Each
10144682 90 mm Each
29 options
Product Code. 10032762
Brand: RETSCH 60.198.050000
Only null left
Specifications
Quantity 1 Each
Pore Size 50 mm
Type Test Sieve
Certifications/Compliance ISO 3310/2
Includes Certificate indicating Compliance
Height (Metric) 50mm
For Use With (Application) For separation, fractioning, particle size determination
Diameter (Metric) Outer 200 mm
Frame Material Stainless Steel
Mesh Material Stainless Steel