Learn More
RETSCH Stainless-Steel Test Sieves, 200 Dia. x 50 mmH, with Perforated Plates with Round Holes and Pore Size 10 mm and Smaller
Product Code. 10174442 Shop All Retsch Products
Change view
Click to view available options
Pore Size:
1 mm
1.18 mm
1.25 mm
1.4 mm
1.6 mm
1.7 mm
2 mm
2.24 mm
2.36 mm
2.5 mm
2.8 mm
3.15 mm
3.35 mm
3.55 mm
4 mm
4.5 mm
4.75 mm
5 mm
5.6 mm
6.3 mm
6.7 mm
7.1 mm
8 mm
9 mm
9.5 mm
10 mm
Unit Size:
Each
Product Code. Pore Size unitSize
10174442 1.25 mm Each
10525173 1.18 mm Each
10266842 1.4 mm Each
10575553 1.6 mm Each
10582673 1.7 mm Each
10032782 10 mm Each
10636703 1 mm Each
10348422 2.5 mm Each
10338422 2.24 mm Each
10798794 2.36 mm Each
10256462 2.8 mm Each
10052962 2 mm Each
10554213 3.15 mm Each
10388562 3.55 mm Each
10022882 4.5 mm Each
10442462 4.75 mm Each
10484952 3.35 mm Each
10494952 4 mm Each
10154972 5.6 mm Each
10398562 5 mm Each
10565363 6.7 mm Each
10686703 6.3 mm Each
10492842 7.1 mm Each
10694583 9 mm Each
10092912 8 mm Each
10779924 9.5 mm Each
26 options
Product Code. 10174442
Brand: RETSCH 60.198.001250
Only null left
Specifications
Quantity 1 Each
Pore Size 1.25 mm
Type Test Sieve
Certifications/Compliance ISO 3310/2
Includes Certificate indicating Compliance
Height (Metric) 50mm
For Use With (Application) For separation, fractioning, particle size determination
Diameter (Metric) Outer 200 mm
Frame Material Stainless Steel
Mesh Material Stainless Steel