Learn More
RETSCH Stainless-Steel Test Sieves, 200 Dia. x 50 mmH, with Perforated Plates with Round Holes and Pore Size 10 mm and Smaller
Product Code. 10092912 Shop All Retsch Products
Change view
Click to view available options
Pore Size:
1 mm
1.18 mm
1.25 mm
1.4 mm
1.6 mm
1.7 mm
2 mm
2.24 mm
2.36 mm
2.5 mm
2.8 mm
3.15 mm
3.35 mm
3.55 mm
4 mm
4.5 mm
4.75 mm
5 mm
5.6 mm
6.3 mm
6.7 mm
7.1 mm
8 mm
9 mm
9.5 mm
10 mm
Unit Size:
Each
Product Code. Pore Size unitSize
10092912 8 mm Each
10525173 1.18 mm Each
10266842 1.4 mm Each
10575553 1.6 mm Each
10174442 1.25 mm Each
10582673 1.7 mm Each
10032782 10 mm Each
10636703 1 mm Each
10348422 2.5 mm Each
10338422 2.24 mm Each
10798794 2.36 mm Each
10256462 2.8 mm Each
10052962 2 mm Each
10554213 3.15 mm Each
10388562 3.55 mm Each
10022882 4.5 mm Each
10442462 4.75 mm Each
10484952 3.35 mm Each
10494952 4 mm Each
10154972 5.6 mm Each
10398562 5 mm Each
10565363 6.7 mm Each
10686703 6.3 mm Each
10492842 7.1 mm Each
10694583 9 mm Each
10779924 9.5 mm Each
26 options
Product Code. 10092912
Brand: RETSCH 60.198.008000
Only null left
Specifications
Quantity 1 Each
Pore Size 8 mm
Type Test Sieve
Certifications/Compliance ISO 3310/2
Includes Certificate indicating Compliance
Height (Metric) 50mm
For Use With (Application) For separation, fractioning, particle size determination
Diameter (Metric) Outer 200 mm
Frame Material Stainless Steel
Mesh Material Stainless Steel