Learn More
- Home
- Products
- Filters and Filtration
- Mechanical Sieves
- RETSCH Stainless-Steel Test Sieves, 200 Dia. x 50 mmH, in Millimeter Pore Sizes with ASTM E 11
RETSCH Stainless-Steel Test Sieves, 200 Dia. x 50 mmH, in Millimeter Pore Sizes with ASTM E 11
Product Code. 10515262 Shop All Retsch Products
Change view
Click to view available options
Pore Size:
1 mm
1.18 mm
1.4 mm
1.7 mm
2 mm
2.36 mm
2.8 mm
3.35 mm
4 mm
4.75 mm
5.6 mm
6.3 mm
6.7 mm
8 mm
9.5 mm
11.2 mm
12.5 mm
13.2 mm
16 mm
19 mm
22.4 mm
25 mm
26.5 mm
31.5 mm
37.5 mm
45 mm
50 mm
53 mm
63 mm
75 mm
90 mm
100 mm
106 mm
125 mm
Unit Size:
Each
| Product Code. | Pore Size | unitSize |
|---|---|---|
| 10515262 | 13.2 mm | Each |
| 10709512 | 1.18 mm | Each |
| 10074751 | 1.4 mm | Each |
| 10288681 | 1.7 mm | Each |
| 10370762 | 100 mm | Each |
| 10065011 | 106 mm | Each |
| 10575062 | 11.2 mm | Each |
| 10781233 | 12.5 mm | Each |
| 10025051 | 125 mm | Each |
| 10289161 | 16 mm | Each |
| 10515452 | 19 mm | Each |
| 10380812 | 1 mm | Each |
| 10218831 | 2.36 mm | Each |
| 10228831 | 2.8 mm | Each |
| 10525452 | 22.4 mm | Each |
| 10268541 | 25 mm | Each |
| 10447961 | 26.5 mm | Each |
| 10330812 | 2 mm | Each |
| 10288871 | 3.35 mm | Each |
| 10576022 | 31.5 mm | Each |
| 10310242 | 4.75 mm | Each |
| 10094901 | 37.5 mm | Each |
| 10136601 | 45 mm | Each |
| 10185971 | 4 mm | Each |
| 10075031 | 5.6 mm | Each |
| 10506222 | 50 mm | Each |
| 10298681 | 53 mm | Each |
| 10498721 | 6.3 mm | Each |
| 10408731 | 6.7 mm | Each |
| 10600272 | 63 mm | Each |
| 10268781 | 75 mm | Each |
| 10574872 | 8 mm | Each |
| 10196211 | 9.5 mm | Each |
| 10278781 | 90 mm | Each |
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Specifications
Specifications
| Quantity | 1 Each |
| Pore Size | 13.2 mm |
| Type | Test Sieve |
| Certifications/Compliance | ASTM E 11 |
| For Use With (Equipment) | Shaker AS 200 |
| Includes | Certificate indicating Compliance |
| Height (Metric) | 50mm |
| For Use With (Application) | For separation, fractioning, particle size determination |
| Diameter (Metric) Outer | 200 mm |
| Frame Material | Stainless Steel |
| Mesh Material | Stainless Steel |
| Show More Show Less |