Learn More
RETSCH Stainless-Steel Test Sieves, 200 Dia. x 50 mmH, in Micrometer Pore with ASTM E 11
Product Code. 10408161 Shop All Retsch Products
Change view
Click to view available options
Pore Size:
20 μm
25 μm
32 μm
38 μm
45 μm
53 μm
63 μm
75 μm
90 μm
106 μm
125 μm
150 μm
180 μm
212 μm
250 μm
300 μm
355 μm
425 μm
500 μm
600 μm
710 μm
850 μm
Unit Size:
Each
Product Code. Pore Size unitSize
10408161 45 μm Each
10300532 106 μm Each
10310532 125 μm Each
10416051 150 μm Each
10620462 180 μm Each
10630282 20 μm Each
10426051 212 μm Each
10228691 250 μm Each
10146411 25 μm Each
10700863 300 μm Each
10299161 32 μm Each
10456431 355 μm Each
10176601 38 μm Each
10015091 425 μm Each
10554112 500 μm Each
10505842 53 μm Each
10208501 63 μm Each
10084921 600 μm Each
10751433 710 μm Each
10268591 75 μm Each
10504692 850 μm Each
10458541 90 μm Each
22 options
Product Code. 10408161
Brand: RETSCH 60.132.000045
Only null left
Specifications
Quantity 1 Each
Pore Size 45 μm
Type Test Sieve
Certifications/Compliance ASTM E 11
For Use With (Equipment) Shaker AS 200
Includes Certificate indicating Compliance
Height (Metric) 50mm
For Use With (Application) For separation, fractioning, particle size determination
Diameter (Metric) Outer 200 mm
Frame Material Stainless Steel
Mesh Material Stainless Steel