Learn More
RETSCH Stainless-Steel Test Sieves, 200 Dia. x 25 mmH, in Micrometer Pore Sizes with ASTM E 11
Shop All Retsch ProductsSpecifications
Specifications
| Quantity | 1 Each |
| Pore Size | 250 μm |
| Type | Test Sieve |
| Certifications/Compliance | ASTM E 11 |
| For Use With (Equipment) | Shaker AS 200 |
| Includes | Certificate indicating Compliance |
| Height (Metric) | 25mm |
| For Use With (Application) | For separation, fractioning, particle size determination |
| Diameter (Metric) Outer | 200 mm |
| Frame Material | Stainless Steel |
| Show More |