Learn More
RETSCH Stainless-Steel Test Sieves, 200 Dia. x 25 mmH, in Micrometer Pore Sizes with ASTM E 11
Product Code. 10024921 Shop All Retsch Products
Change view
Click to view available options
Pore Size:
20 μm
25 μm
32 μm
38 μm
45 μm
53 μm
63 μm
75 μm
90 μm
106 μm
125 μm
150 μm
180 μm
212 μm
250 μm
300 μm
Unit Size:
Each
Product Code. Pore Size unitSize
10024921 106 μm Each
10408541 150 μm Each
10427771 125 μm Each
10690262 180 μm Each
10629911 25 μm Each
10739702 250 μm Each
10095041 20 μm Each
10126601 212 μm Each
10390712 300 μm Each
10771233 32 μm Each
10156311 38 μm Each
10457001 45 μm Each
10166311 53 μm Each
10427581 90 μm Each
10721623 63 μm Each
10106461 75 μm Each
16 options
Product Code. 10024921
Brand: RETSCH 60.123.000106
Only null left
Specifications
Quantity 1 Each
Pore Size 106 μm
Type Test Sieve
Certifications/Compliance ASTM E 11
For Use With (Equipment) Shaker AS 200
Includes Certificate indicating Compliance
Height (Metric) 25mm
For Use With (Application) For separation, fractioning, particle size determination
Diameter (Metric) Outer 200 mm
Frame Material Stainless Steel
Mesh Material Stainless Steel