Learn More
- Home
- Products
- Filters and Filtration
- Mechanical Sieves
- RETSCH Stainless-Steel Test Sieves, 150 mm Dia., in Millimeter Pore Sizes with ISO 3310/1
RETSCH Stainless-Steel Test Sieves, 150 mm Dia., in Millimeter Pore Sizes with ISO 3310/1
Product Code. 10156261 Shop All Retsch Products| Product Code. | Pore Size | unitSize |
|---|---|---|
| 10156261 | 4.75 mm | Each |
| 10770653 | 1.12 mm | Each |
| 10084771 | 1.18 mm | Each |
| 10175971 | 1.25 mm | Each |
| 10310092 | 1.4 mm | Each |
| 10370712 | 11.2 mm | Each |
| 10533912 | 1.7 mm | Each |
| 10467381 | 10 mm | Each |
| 10249161 | 1.8 mm | Each |
| 10523912 | 1.6 mm | Each |
| 10218491 | 12.5 mm | Each |
| 10524102 | 13.2 mm | Each |
| 10534102 | 14 mm | Each |
| 10228491 | 16 mm | Each |
| 10339511 | 18 mm | Each |
| 10450022 | 19 mm | Each |
| 10320042 | 1 mm | Each |
| 10034811 | 2.24 mm | Each |
| 10436041 | 2.36 mm | Each |
| 10721043 | 2.5 mm | Each |
| 10436231 | 2.8 mm | Each |
| 10564482 | 20 mm | Each |
| 10701043 | 22.4 mm | Each |
| 10421962 | 25 mm | Each |
| 10329561 | 26.5 mm | Each |
| 10064841 | 28 mm | Each |
| 10543912 | 2 mm | Each |
| 10741233 | 3.35 mm | Each |
| 10320382 | 3.15 mm | Each |
| 10671202 | 3.55 mm | Each |
| 10074841 | 31.5 mm | Each |
| 10298531 | 35.5 mm | Each |
| 10400032 | 37.5 mm | Each |
| 10044881 | 4.5 mm | Each |
| 10555442 | 40 mm | Each |
| 10524682 | 4 mm | Each |
| 10781803 | 45 mm | Each |
| 10417001 | 5 mm | Each |
| 10701813 | 6.3 mm | Each |
| 10678561 | 5.6 mm | Each |
| 10176401 | 6.7 mm | Each |
| 10320572 | 7.1 mm | Each |
| 10546022 | 9.5 mm | Each |
| 10711813 | 8 mm | Each |
| 10094711 | 9 mm | Each |
Product Code. 10156261
Brand: RETSCH 60.116.004750
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Specifications
| Quantity | 1 Each |
| Pore Size | 4.75 mm |
| Type | Test Sieve |
| Certifications/Compliance | ISO 3310/1 |
| For Use With (Equipment) | Shaker AS 200 |
| Includes | Certificate indicating Compliance |
| Height (Metric) | 40mm |
| For Use With (Application) | For separation, fractioning, particle size determination |
| Diameter (Metric) Outer | 150 mm |
| Frame Material | Stainless Steel |
| Mesh Material | Stainless Steel |
| Show More Show Less |